Learn More
BUCHI FlashPure Empty Solid Loader: Parts
Parts for FlashPure Empty Solid Loaders. FlashPure Solid Loaders are ideal for samples that are not readily soluble in the desired mobile phase when performing flash chromatography. The sample is adsorbed onto a solid phase (silica or Celite) and then added to the empty solid loader tube.
Supplier:  BUCHI 140000178
 View more versions of this product
Catalog No. 05410131
Only null left
Explore available promotions
Description
Description
  • Screw cap makes sealing easy
  • Various sizes for differing sample amounts
  • Choose a size large enough for the sample
  • Insertion Rod allows frit placement and loading of samples smaller than the entire volume
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Specifications
Insertion Rod
1 pc.
80 to120 g
80/120 g Cartridges
Product Suggestions
Product Suggestions