Learn More
Fisherbrand™ Fisherbrand™ Large-Orifice Pipet Tips
Fisherbrand™ Large-Orifice Pipet Tips helps you pipet difficult liquids.
$119.46 - $119.46
Specifications
Tip Style Genomic/Wide Orifice
View More Specs
Products 1
Catalog Number Mfr. No. Format Volume (Metric) Color Length (English) Price Qty  
Catalog Number Mfr. No. Format Volume (Metric) Color Length (English) Price Qty  
02-707-143
missing translation for 'viewDocuments'
N/A Bulk 100 to 1000 ÎĽL Natural 3.4 in.
Pack of 1000 Each for $119.46
Case of 10 Pack for $1,194.60
Only null left
Description
Description
Genomic/Wide Orifice assists in pipetting small volumes of viscous materials.
  • Assist in pipetting small volumes of viscous materials
  • Help eliminate shearing of genomic DNA and fragile cells
Specifications
Specifications
Genomic/Wide Orifice