missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ First Class Filter Pipette Tips, Blue Rack with Lift-off Cover

Description
- Sterile, filtered tips
- Blue rack with lift-off cover
Specifications
Specifications
| Volume (Metric) | 50 to 1200 μL |
| Filtered | Filtered |
| Sterility | Sterile |
| Color | Natural |
| Format | Rack |
| Packaging | 96 tip rack |
| For Use With (Equipment) | Sartorius Biohit Proline Multichannel Electronic |