Learn More
Fisherbrand™ Maxima™ C Plus Pumps, Standard Exhaust Oil Filter Replacement Elements
Replacement filter will help keep the Macima C vacuum pump running smoothly
$245.22 - $423.51
Catalog Number | Mfr. No. | For Use With | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With | Price | Quantity | ||||||
01-257-45
|
Fisherbrand™
0125745 |
Model M8C vacuum pump exhaust filter |
Each for $355.35
|
|
||||||
01-257-6C
|
Fisherbrand™
012576C |
Model M12C and M16C vacuum pump exhaust filter |
Each for $423.51
|
|
||||||
01-257-6A
|
Fisherbrand™
012576A |
Model M2C vacuum pump exhaust filter |
Each for $245.22
|
|
||||||
Description
Replacement filter will help keep the Macima C vacuum pump running smoothly
For exhaust filters supplied with Fisher Scientific Maxima C Plus pumps.
Specifications
Filter Element |