Learn More
Fisher BioReagents™ EZ-Run™ Prestained Rec Protein Ladder, Fisher BioReagents
Comprised of 10 recombinant proteins covalently coupled to a blue chromophore plus 10 and 72kDa reference bands tagged with green and orange dyes, respectively
Supplier:  Fisher BioReagents™ BP3603500
 View more versions of this product
Catalog No. BP3603500
Only null left
Explore available promotions
Description
Description
Comprised of 10 recombinant proteins covalently coupled to a blue chromophore plus 10 and 72kDa reference bands tagged with green and orange dyes, respectively
  • Ladder is ideal for monitoring protein separation during SDS-PAGE, verifying Western blot transfer efficiency and approximating the molecular weight of blotted proteins
  • Supplied in gel loading buffer
The covalently coupled chromophore affects protein mobility; this prestained marker should only be used for approximating the molecular weight of separated proteins.
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Specifications
Blue
Liquid
Monitoring protein separation during SDS-PAGE
10-170 kDa
100 Loadings
Protein Molecular Weight Marker/Ladder
Supplied in gel-loading buffer
500 μL, 100 Loadings
−20°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
This product is manufactured under the license for Strep-tag™ technology covered by U.S. Patent No. 5,506,121; U.K. Patent No. 2272698; and French Patent No. 93 13 066.