missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agilent BioTek Filter Cubes for Imaging Multi-Mode Reader

Description
For use with Imaging Multi-Mode Readers.
Specifications
Specifications
| Type | Filter Cube |
| For Use With (Equipment) | Imaging Multi-Mode Reader |
| Color | Green |
| Label or Dye | GFP |