missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferric Chloride, Colorimetric Solution (CS), 1 mL = 45.0 mg FeCl3.6H2O., 44.5-45.5 mg/mL, Spectrum™ Chemical
F-095, 10025-77-1
Supplier: Spectrum Chemical Mfg Cor F095500MLGL
Description
Spectrum™ Chemical Ferric Chloride, Colorimetric Solution (CS), 1 ml = 45.0 mg FeCl3.6H2O. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
10025-77-1,7732-18-5 | |
500 mL | |
MFCD00149712 | |
NQXWGWZJXJUMQB-UHFFFAOYSA-K | |
iron(3+) hexahydrate trichloride | |
44.5-45.5 mg/mL |
4.5%,94.5% | |
Cl3FeH12O6 | |
UN2582 | |
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] | |
270.29 | |
Amber Glass Bottle |
Chemical Identifiers
10025-77-1 | |
270.29 | |
NQXWGWZJXJUMQB-UHFFFAOYSA-K | |
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] |
Cl3FeH12O6 | |
MFCD00149712 | |
iron(3+) hexahydrate trichloride |