Learn More
Thermo Scientific™ Lambda DNA/EcoRI Marker
Perform sizing and approximate quantification of large double-stranded DNA fragments in agarose gels.
Supplier: Thermo Scientific™ SM0281
Description
Lambda DNA is digested to completion with the appropriate Thermo Scientitific restriction enzyme(s) and purified and dissolved in storage buffer. DNA fragments contain various sticky ends depending on the restriction enzyme used for preparation of the marker.
marker can be labeled radioactively with T4 Polynucleotide Kinase. Alternatively, it can be labeled radioactively or non-radioactively with Klenow Fragment or Klenow Fragment, exo- with the fill-in reaction (see Recources for Protocols).
Highlights
- Sizing and approximate quantification of large DNA fragments
- Sharp bands
- Supplied with loading dye for sample DNA
- Environmental benefits include:
- Recyclable
Specifications
6X | |
Xylene Cyanol FF, Bromophenol Blue | |
No | |
Sustainable packaging |
Agarose | |
DNA/ECORI Marker | |
Agarose Gels | |
5 x 50 μg |
For Research Use Only. Not for use in diagnostic procedures.