Learn More
Thermo Scientific™ Lambda DNA/EcoRI plus HindIII Marker
GREENER_CHOICE
Perform sizing and approximate quantification of large double-stranded DNA fragments in agarose gels with this ladder that consists of 13 sharp bands.
$148.00 - $564.00
Specifications
Gel Type Agarose
Product Type DNA/ECORI Plus Hind III Marker
Ready to Load No
Gel Compatibility Agarose Gels
Green Features Sustainable packaging
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
FERSM0191
GREENER_CHOICE
View Documents
Thermo Scientific™
SM0191
5 x 50 μg N/A
Only null left
FERSM0192
GREENER_CHOICE
View Documents
Thermo Scientific™
SM0192
25 x 50 μg N/A
Only null left
Description
Description
Lambda DNA/EcoRI+HindIII Marker is recommended for sizing of linear double-stranded large DNA fragments in agarose gels. Lambda DNA is digested to completion with the appropriate Thermo Scientitfic restriction enzyme(s) and purified and dissolved in storage buffer. DNA fragments contain various sticky ends depending on the restriction enzyme used for preparation of the marker.
Lambda DNA/EcoRI+HindIII Marker can be labeled radioactively with T4 Polynucleotide Kinase. Alternatively, and it can be labeled radioactively or non-radioactively with Klenow Fragment or Klenow Fragment, exo- with the fill-in reaction.
Highlights
  • Sizing and approximate quantification of large DNA fragments
  • Sharp bands
  • Supplied with loading dye for sample DNA
Specifications
Specifications
Agarose
No
Sustainable packaging
DNA/ECORI Plus Hind III Marker
Agarose Gels
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
For Research Use Only. Not for use in diagnostic procedures.