missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Exo-Resistant Random Primer

Thermo Scientific Exonuclease-Resistant Random Primer is a mixture of single-stranded random oligonucleotides used for highly efficient random priming of various DNA synthesis reactions.

Supplier:  Thermo Scientific™ SO181

Catalog No. FERSO181


Only null left
Explore available promotions

Description

Description

Thermo Scientific Exonuclease-Resistant Random Primer is a mixture of single-stranded random oligonucleotides used for highly efficient random priming of various DNA synthesis reactions. This primer has two 3'-terminal phosphorothioate (PTO) modifications that are resistant to the 3'→5' exonuclease activity of proofreading DNA polymerases, such as Klenow Fragment and phi29 DNA Polymerase. It also has 5'- and 3'-hydroxyl ends. The Exo-Resistant Random Primer is supplied as a ready-to-use, 20X concentrated aqueous solution.

Features
• 3'-terminal PTO modifications protect from 3'→5' exonuclease degradation and ensure efficient priming of DNA synthesis
• Product delivered in a ready-to-use format

Applications
• Strand displacement amplification of genomic DNA, plasmids, and phage DNA
• DNA labeling by random priming

Greener Choice Claims
  • Environmental benefits include:
    • Recyclable
Specifications

Specifications

Primer
Dry Ice
Random
DNA Labeling, Strand Displacement Amplification
20X Exo-Resistant Random Primer (100 μL)

Store at –20°C.
500 μM
100 μL
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.