missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ethylethiomorpholine-3-carboxylate hydrochloride, 90%, Thermo Scientificâ„¢
Chemical Identifiers
CAS | 159381-07-4 |
---|---|
Molecular Formula | C7H14ClNO2S |
Molecular Weight (g/mol) | 211.704 |
MDL Number | MFCD00107968 |
InChI Key | QQRCEZBURAQLBQ-UHFFFAOYSA-N |
Synonym | ethyl thiomorpholine-3-carboxylate hydrochloride, thiomorpholine-3-carboxylic acid ethyl ester hydrochloride, thiomorpholine-3-carboxylic acid ethyl ester hcl, thiomorpholine-3-carboxylicacidethylesterhydrochloride, ethyl thiomorpholine-3-carboxylate hcl, ethylthiomorpholine-3-carboxylatehydrochloride, 3-ethoxycarbonyl thiomorpholine hydrochloride, ethyl 1,4-thiazinane-3-carboxylate hydrochloride, ethyl 1,4-thiazaperhydroine-3-carboxylate, chloride, thiomorpholine-3-carboxylic acid ethyl esterhydrochloride |
PubChem CID | 17433447 |
IUPAC Name | ethyl thiomorpholine-3-carboxylate;hydrochloride |
SMILES | CCOC(=O)C1CSCCN1.Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
CC27673DA
|
Thermo Scientific Chemicals
CC27673DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC27673DE
|
Thermo Scientific Chemicals
CC27673DE |
5g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
159381-07-4 | |
211.704 | |
QQRCEZBURAQLBQ-UHFFFAOYSA-N | |
17433447 | |
CCOC(=O)C1CSCCN1.Cl |
C7H14ClNO2S | |
MFCD00107968 | |
ethyl thiomorpholine-3-carboxylate hydrochloride, thiomorpholine-3-carboxylic acid ethyl ester hydrochloride, thiomorpholine-3-carboxylic acid ethyl ester hcl, thiomorpholine-3-carboxylicacidethylesterhydrochloride, ethyl thiomorpholine-3-carboxylate hcl, ethylthiomorpholine-3-carboxylatehydrochloride, 3-ethoxycarbonyl thiomorpholine hydrochloride, ethyl 1,4-thiazinane-3-carboxylate hydrochloride, ethyl 1,4-thiazaperhydroine-3-carboxylate, chloride, thiomorpholine-3-carboxylic acid ethyl esterhydrochloride | |
ethyl thiomorpholine-3-carboxylate;hydrochloride |
Specifications
159381-07-4 | |
100.0 | |
Amber Glass Bottle | |
MFCD00107968 | |
ethyl thiomorpholine-3-carboxylate hydrochloride, thiomorpholine-3-carboxylic acid ethyl ester hydrochloride, thiomorpholine-3-carboxylic acid ethyl ester hcl, thiomorpholine-3-carboxylicacidethylesterhydrochloride, ethyl thiomorpholine-3-carboxylate hcl, ethylthiomorpholine-3-carboxylatehydrochloride, 3-ethoxycarbonyl thiomorpholine hydrochloride, ethyl 1,4-thiazinane-3-carboxylate hydrochloride, ethyl 1,4-thiazaperhydroine-3-carboxylate, chloride, thiomorpholine-3-carboxylic acid ethyl esterhydrochloride | |
CCOC(=O)C1CSCCN1.Cl | |
211.704 | |
211.71 | |
ethylethiomorpholine-3-carboxylate hydrochloride |
89.5 | |
90% | |
C7H14ClNO2S | |
1g | |
QQRCEZBURAQLBQ-UHFFFAOYSA-N | |
ethyl thiomorpholine-3-carboxylate;hydrochloride | |
17433447 | |
90% |