Learn More
Ethyl propiolate, 99%
CAS: 623-47-2 | C5H6O2 | 98.10 g/mol
Supplier: Thermo Scientific Chemicals 118900250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Environmental benefits include:
- Renewable Energy
Specifications
Ethyl propiolate | |
Colorless to Yellow | |
120°C | |
Authentic | |
Glass bottle | |
1.4110 to 1.413 | |
MFCD00009184 | |
02, 477 | |
15, 3898 | |
Solubility in water: miscible with water. Other solubilities: soluble in most organic solvents | |
CCOC(=O)C#C | |
98.10 | |
CHEBI:51740 | |
99% |
623-47-2 | |
0.9680g/mL | |
23°C | |
98.5% min. (GC) | |
C5H6O2 | |
HC∽CCO2C2H5 | |
25 g | |
0.968 | |
ethyl propiolate, ethyl acetylenecarboxylate, 2-propynoic acid, ethyl ester, propiolic acid ethyl ester, ethyl propynoate, ethoxycarbonyl acetylene, ethyl 2-propynoate, propiolic acid, ethyl ester, unii-w235g5u52s, propynoic acid ethyl ester | |
FMVJYQGSRWVMQV-UHFFFAOYSA-N | |
ethyl prop-2-ynoate | |
12182 | |
98.1 | |
Liquid |
Chemical Identifiers
623-47-2 | |
98.10 | |
FMVJYQGSRWVMQV-UHFFFAOYSA-N | |
12182 | |
ethyl prop-2-ynoate |
C5H6O2 | |
MFCD00009184 | |
ethyl propiolate, ethyl acetylenecarboxylate, 2-propynoic acid, ethyl ester, propiolic acid ethyl ester, ethyl propynoate, ethoxycarbonyl acetylene, ethyl 2-propynoate, propiolic acid, ethyl ester, unii-w235g5u52s, propynoic acid ethyl esterShow More | |
CHEBI:51740 | |
CCOC(=O)C#C |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 210-795-8
RUO – Research Use Only