missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene Glycol Dibutyl Ether 98.0+%, TCI Americaâ„¢
$103.94 - $655.75
Chemical Identifiers
CAS | 112-48-1 |
---|---|
Molecular Formula | C10H22O2 |
Molecular Weight (g/mol) | 174.28 |
MDL Number | MFCD00048785 |
InChI Key | GDXHBFHOEYVPED-UHFFFAOYSA-N |
Synonym | 1,2-Dibutoxyethane, Dibutyl Cellosolve, Dibutyl Glycol |
PubChem CID | 8188 |
IUPAC Name | 1-(2-butoxyethoxy)butane |
SMILES | CCCCOCCOCCCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
E021725ML
|
TCI America
E021725ML |
25 mL |
N/A
|
|
||||||
E0217500ML
|
TCI America
E0217500ML |
500 mL |
N/A
|
|
||||||
Chemical Identifiers
112-48-1 | |
174.28 | |
GDXHBFHOEYVPED-UHFFFAOYSA-N | |
8188 | |
CCCCOCCOCCCC |
C10H22O2 | |
MFCD00048785 | |
1,2-Dibutoxyethane, Dibutyl Cellosolve, Dibutyl Glycol | |
1-(2-butoxyethoxy)butane |
Specifications
112-48-1 | |
Colorless | |
C10H22O2 | |
25 mL | |
GDXHBFHOEYVPED-UHFFFAOYSA-N | |
1-(2-butoxyethoxy)butane | |
8188 | |
≥98.0% (GC) | |
Ethylene Glycol Dibutyl Ether |
-69°C | |
202°C | |
MFCD00048785 | |
1,2-Dibutoxyethane, Dibutyl Cellosolve, Dibutyl Glycol | |
CCCCOCCOCCCC | |
174.28 | |
174.28 | |
Liquid |
Safety and Handling
EINECSNumber : (2)-0420&(7)-1321
RTECSNumber : KH9450000
TSCA : No