Learn More
Ethyl caproate, 99+%
CAS: 123-66-0 | C8H16O2 | 144.21 g/mol
Supplier: Thermo Scientific Chemicals 164995000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Environmental benefits include:
- Renewable Energy
Specifications
Ethyl caproate | |
1mg KOH/g max. | |
Colorless | |
168°C | |
Authentic | |
Glass bottle | |
1.4060 to 1.409 | |
MFCD00009511 | |
02, 323 | |
15, 3832 | |
Solubility in water: insoluble. Other solubilities: insoluble in glycerol,soluble in alcohol and ether | |
CCCCCC(=O)OCC | |
144.21 | |
CHEBI:86055 | |
99+% |
123-66-0 | |
67°C | |
0.8690g/mL | |
54°C | |
99% min. (GC) | |
C8H16O2 | |
CH3(CH2)4CO2C2H5 | |
500 mL | |
0.869 | |
ethyl caproate, ethyl hexoate, hexanoic acid, ethyl ester, hexanoic acid ethyl ester, capronic ether absolute, caproic acid ethyl ester, ethyl n-hexanoate, ethyl butyl acetate, caproate, acetic acid, butyl-, ethyl ester | |
SHZIWNPUGXLXDT-UHFFFAOYSA-N | |
ethyl hexanoate | |
31265 | |
144.21 | |
Liquid |
Chemical Identifiers
123-66-0 | |
144.21 | |
SHZIWNPUGXLXDT-UHFFFAOYSA-N | |
31265 | |
ethyl hexanoate |
C8H16O2 | |
MFCD00009511 | |
ethyl caproate, ethyl hexoate, hexanoic acid, ethyl ester, hexanoic acid ethyl ester, capronic ether absolute, caproic acid ethyl ester, ethyl n-hexanoate, ethyl butyl acetate, caproate, acetic acid, butyl-, ethyl esterShow More | |
CHEBI:86055 | |
CCCCCC(=O)OCC |
Safety and Handling
GHS H Statement:
Flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 204-640-3
RUO – Research Use Only