missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ethyle6-iodo-4-oxo-4H-chromene-2-carboxylate, 97%, Thermo Scientific™

Chemical Identifiers
CAS | 35204-44-5 |
---|---|
Molecular Formula | C12H9IO4 |
Molecular Weight (g/mol) | 344.10 |
MDL Number | MFCD00100378 |
InChI Key | HUJFMOAXSPSGDU-UHFFFAOYSA-N |
Synonym | ethyl 6-iodo-4-oxo-4h-chromene-2-carboxylate, 4h-1-benzopyran-2-carboxylicacid, 6-iodo-4-oxo-, ethyl ester, maybridge1_002002, ethyl-6-iodo-4-oxo-4h-chromene-2-carboxylate, 4-oxo-6-iodo-4h-1-benzopyran-2-carboxylic acid ethyl ester |
PubChem CID | 2775235 |
IUPAC Name | ethyl 6-iodo-4-oxo-4H-chromene-2-carboxylate |
SMILES | CCOC(=O)C1=CC(=O)C2=CC(I)=CC=C2O1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
BTB10203DA
|
Thermo Scientific Chemicals
BTB10203DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
BTB10203EA
|
Thermo Scientific Chemicals
BTB10203EA |
10g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
35204-44-5 | |
344.10 | |
HUJFMOAXSPSGDU-UHFFFAOYSA-N | |
2775235 | |
CCOC(=O)C1=CC(=O)C2=CC(I)=CC=C2O1 |
C12H9IO4 | |
MFCD00100378 | |
ethyl 6-iodo-4-oxo-4h-chromene-2-carboxylate, 4h-1-benzopyran-2-carboxylicacid, 6-iodo-4-oxo-, ethyl ester, maybridge1_002002, ethyl-6-iodo-4-oxo-4h-chromene-2-carboxylate, 4-oxo-6-iodo-4h-1-benzopyran-2-carboxylic acid ethyl ester | |
ethyl 6-iodo-4-oxo-4H-chromene-2-carboxylate |
Specifications
35204-44-5 | |
100.0 | |
Amber Glass Bottle | |
MFCD00100378 | |
ethyl 6-iodo-4-oxo-4h-chromene-2-carboxylate, 4h-1-benzopyran-2-carboxylicacid, 6-iodo-4-oxo-, ethyl ester, maybridge1_002002, ethyl-6-iodo-4-oxo-4h-chromene-2-carboxylate, 4-oxo-6-iodo-4h-1-benzopyran-2-carboxylic acid ethyl ester | |
CCOC(=O)C1=CC(=O)C2=CC(I)=CC=C2O1 | |
344.10 | |
344.1 | |
Ethyl 6-iodo-4-oxo-4H-chromene-2-carboxylate |
96.5 | |
97% | |
C12H9IO4 | |
1g | |
HUJFMOAXSPSGDU-UHFFFAOYSA-N | |
ethyl 6-iodo-4-oxo-4H-chromene-2-carboxylate | |
2775235 | |
97% |