Learn More
Ethyl 2-cyclohexanoneacetate, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 118320250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Environmental benefits include:
- Renewable Energy
Specifications
Ethyl 2-cyclohexanoneacetate | |
1.0200g/mL | |
Authentic | |
Glass bottle | |
1.4570 to 1.4590 (20°C, 589nm) | |
MFCD00001638 | |
1.02 | |
ZZWSNYNCRUZSPR-UHFFFAOYSA-N | |
ethyl 2-(2-oxocyclohexyl)acetate | |
91229 | |
97% |
24731-17-7 | |
>110°C | |
96% min. (GC) | |
C10H16O3 | |
(O=)C6H9CH2CO2C2H5 | |
25g | |
ethyl 2-2-oxocyclohexyl acetate, ethyl 2-cyclohexanoneacetate, ethyl 2-oxocyclohexaneacetate, ethyl 2-oxocyclohexyl acetate, ethyl 2-cyclohexanone acetate, ethyl-2-cyclohexanone acetate, ethyl 2-oxocyclohexylacetate, cyclohexaneacetic acid, 2-oxo-, ethyl ester, ethyl 2-oxo-cyclohexyl-acetate | |
CCOC(=O)CC1CCCCC1=O | |
184.23 | |
184.23 |
Chemical Identifiers
24731-17-7 | |
184.23 | |
ZZWSNYNCRUZSPR-UHFFFAOYSA-N | |
91229 | |
CCOC(=O)CC1CCCCC1=O |
C10H16O3 | |
MFCD00001638 | |
ethyl 2-2-oxocyclohexyl acetate, ethyl 2-cyclohexanoneacetate, ethyl 2-oxocyclohexaneacetate, ethyl 2-oxocyclohexyl acetate, ethyl 2-cyclohexanone acetate, ethyl-2-cyclohexanone acetate, ethyl 2-oxocyclohexylacetate, cyclohexaneacetic acid, 2-oxo-, ethyl ester, ethyl 2-oxo-cyclohexyl-acetateShow More | |
ethyl 2-(2-oxocyclohexyl)acetate |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 246-435-1
RUO â Research Use Only