Learn More
Bioassay Systems EnzyChrom™ Ethanol Assay Kit. For quantitative determination of ethanol and evaluation of drug effects on alcohol metabolism.
Supplier: Bioassay Systems ECET100
For quantitative determination of ethanol and evaluation of drug effects on alcohol metabolism. Key Features: Sensitive and accurate. Detection limit 0.0008 vol % (140 µM or 8 ppm), linearity up to 0.1% ethanol in 96-well plate assay. Convenient. The procedure involves adding working reagent, incubating for 30 min and stopping reaction. No 37°C heater is needed. High-throughput. Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day. Method: OD565nm (Enzymatic). Samples: Serum, plasma, urine, saliva samples etc. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: 0.0008%.