Learn More
MilliporeSigma™ Vent Filters for Pure Water Storage Reservoirs
For use with Pure Water Reservoirs
Supplier:  MilliporeSigma™ TANKMPK01
 View more versions of this product
Catalog No. TANKMPK01
Only null left
Explore available promotions
Description
Description
For use with Pure Water Reservoirs
  • Polyethylene vent filters protect purified water stored in Pure Water Reservoirs when used with low-volume Elix™ or RiOs™ Water Purification Systems from ingress of airborne microorganisms
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Specifications
Polyethylene
Elix™ 3/5/10 system
Vent Filter
Product Suggestions
Product Suggestions