missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Wedge Model

Demonstrates how a wedge can increase mechanical advantage.

Supplier:  Eisco™ WDMS14

Catalog No. S07822


Only null left
Explore available promotions

Includes:

Activity guide


Description

Description

  • Demonstrates principles of physics
  • Solid wood construction is ideal for hands-on classroom use
Greener Choice Claims
  • Environmental benefits include:
    • Source Reduction
Specifications

Specifications

Model, Wedge
Wood
Demonstrate how a wedge can increase mechanical advantage
Activity guide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions