Learn More
DuPont™ Tyvek™ IsoClean™ Series 253 Bound Seam Coveralls, Bulk Packaged
Tyvek coveralls provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash. Bound seams reduce the potential for particle penetration.
Supplier:  DuPont™ IC253BWHLG00250B
 View more versions of this product
Catalog No. 17-987-993
Only null left
Explore available promotions
Alert:
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
Description
  • For use in controlled environments with high standards for particle and microbiological contamination control:
    • Pharmaceutical
    • Biotech
    • Medical Device
    • Electronics
  • Light and durable
  • Spun polyethylene creates barrier to particles, micro-organisms, and non-hazardous liquids
  • Lowest level particle shedding
  • Bound seams and neck
  • Covered elastic wrists and ankles with dolman sleeves
  • Zipper closure
  • Should not be worn around open flame, heat source, sparks or ignition sources
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Show More
Product Suggestions
Product Suggestions