missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ NexGen™ Lab Coats
Limited-use general purpose lab coats are suitable for protection against non-hazardous particles, light liquid splash, and aerosols in many industrial applications.
Supplier: DuPont™ NG212SWHLG003000
Description
- ProShield 60 lab coat
- Open wrists
- Extends below hip
- Front snap closure
- Pockets, serged seams
- NexGen is suited for nontoxic liquid and spray hazards
- Lab coats are generously cut for ease of movement
Specifications
General Purpose Lab Coat | |
Unisex | |
White | |
Open | |
Non-sterile | |
2 | |
NG212S | |
36.25 to 39.75 in. | |
33.25 in. |
Large | |
ProShield NexGen | |
Snaps | |
Limited-use | |
Light Liquid Splash | |
Lay Down | |
Serged | |
Automotive Refinishing, Waste Cleanup, Remediation | |
84.45 cm |