missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mopec Double Faucet Autopsy Scrub Sink

Essential product for any anatomy lab providing a specific place to wash hands before and after an autopsy or any handling of infectious material. Mopec™ Double Faucet Autopsy Scrub Sink has no parting seams which can potentially collect bacteria.

Supplier:  Mopec CH200

Catalog No. 22444043

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

  • Stainless steel scrub sink designed with a 12 in. backsplash
  • Hot and cold foot pedal valves with two gooseneck faucet

Warranty

1 Year Warranty

Specifications

Specifications

Autopsy and Embalming Sink
19 in.
48 in.
58.5 cm
Stainless Steel
48.7 cm
23 in.
121 cm
SDS
Documents

Documents

Promotions

Promotions