Learn More
DL-Prolinol, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6232214
Description
DL-Prolinol is used as asymmetric catalysts in proline organ catalysis reactions, and it is also used in many pharmaceutical, biotechnological applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDL-Prolinol is used as asymmetric catalysts in proline organ catalysis reactions, and it is also used in many pharmaceutical, biotechnological applications.
Solubility
Soluble in ethanol.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
DL-Prolinol | |
1.484 | |
25 g | |
MFCD00601073 | |
Soluble in ethanol. | |
C1CC(NC1)CO | |
101.149 | |
CHEBI:59652 | |
98% |
498-63-5 | |
76°C (2mmHg) | |
C5H11NO | |
2-hydroxymethyl pyrrolidine, 2-pyrrolidinemethanol, prolinol, pyrrolidin-2-yl-methanol, dl-prolinol, 2-hydroxymethylpyrrolidine, pyrrolidin-2-yl methanol, 2-pyrrolidinylmethanol, pyrrolidine-2-methanol, 2-pyrrolidinylmethanol # | |
HVVNJUAVDAZWCB-UHFFFAOYSA-N | |
pyrrolidin-2-ylmethanol | |
90908 | |
101.15 |
Chemical Identifiers
498-63-5 | |
101.149 | |
HVVNJUAVDAZWCB-UHFFFAOYSA-N | |
90908 | |
pyrrolidin-2-ylmethanol |
C5H11NO | |
MFCD00601073 | |
2-hydroxymethyl pyrrolidine, 2-pyrrolidinemethanol, prolinol, pyrrolidin-2-yl-methanol, dl-prolinol, 2-hydroxymethylpyrrolidine, pyrrolidin-2-yl methanol, 2-pyrrolidinylmethanol, pyrrolidine-2-methanol, 2-pyrrolidinylmethanol # | |
CHEBI:59652 | |
C1CC(NC1)CO |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only