Learn More
DL-Mandelic acid, 99+%
CAS: 90-64-2 | C8H8O3 | 152.149 g/mol
Supplier: Thermo Scientific Chemicals 125310010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Environmental benefits include:
- Recyclable
Specifications
DL-Mandelic acid | |
117°C to 121°C | |
White | |
C8H8O3 | |
mandelic acid, dl-mandelic acid, phenylglycolic acid, amygdalic acid, almond acid, uromaline, p-mandelic acid, racemic mandelic acid, paramandelic acid, rs-mandelic acid | |
C1=CC=C(C=C1)C(C(=O)O)O | |
152.149 | |
CHEBI:35825 | |
≥99.0% (Acid) | |
Authentic | |
Crystals or Crystalline Powder |
99+% | |
90-64-2 | |
99+% | |
C6H5CH(OH)CO2H | |
IWYDHOAUDWTVEP-UHFFFAOYSA-N | |
2-hydroxy-2-phenylacetic acid | |
1292 | |
152.15 | |
1 kg | |
Plastic bottle |
Chemical Identifiers
90-64-2 | |
152.149 | |
mandelic acid, dl-mandelic acid, phenylglycolic acid, amygdalic acid, almond acid, uromaline, p-mandelic acid, racemic mandelic acid, paramandelic acid, rs-mandelic acid | |
CHEBI:35825 | |
C1=CC=C(C=C1)C(C(=O)O)O |
C8H8O3 | |
IWYDHOAUDWTVEP-UHFFFAOYSA-N | |
1292 | |
2-hydroxy-2-phenylacetic acid |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
EINECSNumber : 202-007-6
RTECSNumber : OO6300000
TSCA : TSCA
RUO – Research Use Only