missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ DL-Isoserine, 98%

Chemical Identifiers
CAS | 565-71-9 |
---|---|
Molecular Formula | C3H7NO3 |
Molecular Weight (g/mol) | 105.09 |
MDL Number | MFCD00008138 |
InChI Key | BMYNFMYTOJXKLE-UHFFFAOYSA-N |
Synonym | dl-isoserine, isoserine, 3-amino-2-hydroxypropionic acid, propanoic acid, 3-amino-2-hydroxy, 2-hydroxy-3-aminopropanoic acid, h-dl-isoser-oh, inverted exclamation marka-3-amino-2-hydroxypropionic acid, rs-isoserine, pubchem23947Show More |
PubChem CID | 11267 |
IUPAC Name | 3-amino-2-hydroxypropanoic acid |
SMILES | C(C(C(=O)O)O)N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC279672500
|
Thermo Scientific™
279672500 |
250mg | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC279670010
|
Thermo Scientific™
279670010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
565-71-9 | |
105.09 | |
BMYNFMYTOJXKLE-UHFFFAOYSA-N | |
11267 | |
C(C(C(=O)O)O)N |
C3H7NO3 | |
MFCD00008138 | |
dl-isoserine, isoserine, 3-amino-2-hydroxypropionic acid, propanoic acid, 3-amino-2-hydroxy, 2-hydroxy-3-aminopropanoic acid, h-dl-isoser-oh, inverted exclamation marka-3-amino-2-hydroxypropionic acid, rs-isoserine, pubchem23947Show More | |
3-amino-2-hydroxypropanoic acid |
Specifications
565-71-9 | |
98% | |
C3H7NO3 | |
MFCD00008138 | |
dl-isoserine, isoserine, 3-amino-2-hydroxypropionic acid, propanoic acid, 3-amino-2-hydroxy, 2-hydroxy-3-aminopropanoic acid, h-dl-isoser-oh, inverted exclamation marka-3-amino-2-hydroxypropionic acid, rs-isoserine, pubchem23947 | |
C(C(C(=O)O)O)N | |
105.09 | |
105.09 | |
DL-Isoserine |
Authentic | |
Glass bottle | |
NH2CH3CH(OH)COOH | |
250mg | |
BMYNFMYTOJXKLE-UHFFFAOYSA-N | |
3-amino-2-hydroxypropanoic acid | |
11267 | |
98% |
RUO â Research Use Only