Learn More
DL-Isoborneol, 93%, tech., Thermo Scientific™
Supplier: Thermo Scientific Chemicals 122495000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Environmental benefits include:
- Renewable Energy
Specifications
DL-Isoborneol | |
124-76-5 | |
Authentic | |
Plastic bottle | |
MFCD00074821 | |
06, II, 80 | |
dl-isoborneol | |
DTGKSKDOIYIVQL-SZBHIRRCSA-N | |
(1R,3R)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-ol | |
126961757 | |
93% |
tech., 93% | |
74°C | |
91% min. (GC) | |
C10H18O | |
500g | |
15, 5173 | |
Solubility in water: insoluble in water. Other solubilities: readily soluble in alcohol,chloroform and ether | |
CC1(C2CCC1(C(C2)O)C)C | |
154.25 | |
154.25 | |
Technical |
Safety and Handling
GHS H Statement
Flammable solid.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
GHS Signal Word: Danger
EINECSNumber : 204-712-4
RTECSNumber : NP7300000
TSCA : TSCA
RUO â Research Use Only