Learn More
Honeywell™ DC300 Disposable N95 Respirator, 20/Box
NIOSH approved respirator is designed to comfortably fit most facial shapes while providing N95 respiratory protection.
Supplier:  Honeywell™ DC300N95
Catalog No. 19-085-584
Only null left
Explore available promotions
Description
Description
  • Offered in a universal size
  • The form-fitted mask contours with the natural shape of the face, minimizing pressure points
  • The molded nose bridge reduces adjustments needed to achieve proper fit and provide a secure seal
Greener Choice Claims
  • Environmental benefits include:
    • Source Reduction
Specifications
Specifications
N - Not Resistant to Oil
Cup/Molded
N95
One Size
95%
DC300N95
20/Box
Woven Head Straps
NIOSH 42 CFR 84, N95
Single-use
General Purpose Particulate Respirator
No
Yes
Molded Nose Bridge
Non-sterile
No
Product Suggestions
Product Suggestions