missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Disposable Frocks
White 58 gsm polypropylene and polyethylene frocks that create a barrier against particles, micro-organisms and nonhazardous liquids.
Supplier: Fisherbrand™ 12893002D
Description
- Front zipper closure
- Elastic cuffs
- Snap-closure collar
- Knee-length coverage
Greener Choice Claims
- Environmental benefits include:
- Source Reduction
Specifications
Knee-length Frock | |
Polypropylene, Polyethylene | |
Single-use | |
Zipper | |
58 gsm | |
110 cm | |
24 in. |
Large | |
White | |
Non-sterile | |
Elastic | |
43 in. | |
60 cm |