missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Disposable Frocks
White 58 gsm polypropylene and polyethylene frocks that create a barrier against particles, micro-organisms and nonhazardous liquids.
Supplier: Fisherbrand™ 12893002A
Description
- Front zipper closure
- Elastic cuffs
- Snap-closure collar
- Knee-length coverage
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
Knee-length Frock | |
Polypropylene, Polyethylene | |
Single-use | |
Zipper | |
58 gsm | |
95 cm | |
19 in. |
X-Small | |
White | |
Non-sterile | |
Elastic | |
37 in. | |
48 cm |