missing translation for 'onlineSavingsMsg'
Learn More

Diethylvinylphosphonate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01036809

 View more versions of this product

Catalog No. AA1036809


Only null left
Explore available promotions

Description

Description

Diethylvinylphosphonate is used in the preparation of diethyl N-alkyl-2-aminoethylphosphonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Greener Choice Claims
  • Environmental benefits include:
    • Free-of a substance that causes environmental harm
    • Ozone-Safe or Safe for the Ozone Layer
    • Renewable Energy
    • Non-Toxic
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, LIQUID, TOXIC, N.O.S.

EINECSNumber : 211-663-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only