Learn More
Diethylene glycol diethyl ether, HPLC Grade, 99+%
CAS: 112-36-7 | C8H18O3 | 162.229 g/mol
Supplier: Thermo Scientific Chemicals 043464AK
Description
Diethylene glycol diethyl ether is used to study the activities of enzymes in aqueous organic mixtures.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Diethylene glycol diethyl ether | |
112-36-7 | |
188°C to 190°C | |
71°C (160°F) | |
1.412 | |
MFCD00009254 | |
14,3118 | |
Fully miscible in water. | |
CCOCCOCCOCC | |
162.229 | |
CHEBI:44664 | |
≥99% |
-44°C | |
0.909 g/mL | |
250 mL | |
Liquid | |
C8H18O3 | |
1699259 | |
diethylene glycol diethyl ether, 2-ethoxyethyl ether, diethyl carbitol, ethyl diglyme, 1-ethoxy-2-2-ethoxyethoxy ethane, bis 2-ethoxyethyl ether, diethyldiethylene glycol, 3,6,9-trioxaundecane, degdee, ether, bis 2-ethoxyethyl | |
RRQYJINTUHWNHW-UHFFFAOYSA-N | |
1-ethoxy-2-(2-ethoxyethoxy)ethane | |
8179 | |
162.23 | |
HPLC |
Chemical Identifiers
112-36-7 | |
162.229 | |
RRQYJINTUHWNHW-UHFFFAOYSA-N | |
8179 | |
1-ethoxy-2-(2-ethoxyethoxy)ethane |
C8H18O3 | |
MFCD00009254 | |
diethylene glycol diethyl ether, 2-ethoxyethyl ether, diethyl carbitol, ethyl diglyme, 1-ethoxy-2-2-ethoxyethoxy ethane, bis 2-ethoxyethyl ether, diethyldiethylene glycol, 3,6,9-trioxaundecane, degdee, ether, bis 2-ethoxyethylShow More | |
CHEBI:44664 | |
CCOCCOCCOCC |
Safety and Handling
GHS H Statement
H319-H227
Causes serious eye irritation.
Combustible liquid.
P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
H227-H315-H319
EINECSNumber : 203-963-7
RTECSNumber : KN3160000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only