Learn More
Diethylbenzene, Mixture of o-, m- and p-isomers, Sum Min. 95%
CAS: 25340-17-4 | C10H14 | 134.22 g/mol
$120.74 - $246.99
Chemical Identifiers
CAS | 25340-17-4 |
---|---|
Molecular Formula | C10H14 |
Molecular Weight (g/mol) | 134.22 |
MDL Number | MFCD00792900 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC114160010
|
Thermo Scientific Chemicals
114160010 |
1 L | Glass bottle |
Each for $120.74
|
|
|||||
AC114160025
|
Thermo Scientific Chemicals
114160025 |
2.5 L | Glass bottle |
Each for $246.99
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
25340-17-4 | |
C10H14 | |
134.22 | |
MFCD00792900 |
Specifications
25340-17-4 | |
Colorless to Yellow | |
180.0°C to 182.0°C | |
Authentic | |
C10H14 | |
MFCD00792900 | |
05,IV,1067 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol, benzene, ether and carbon, tetrachloride | |
0.89 mPa.s (20°C) | |
95% | |
Diethylbenzene |
-73.0°C | |
0.8690g/mL | |
56°C | |
Glass bottle | |
1.4940 to 1.4960 | |
1 L | |
0.869 | |
134.22 | |
134.22 | |
Liquid |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.<b
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do
GHS Signal Word: Danger
EINECSNumber : 246-874-9
RTECSNumber : CZ5600000
TSCA : TSCA
RUO – Research Use Only