missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dibutyl sebacate, 93%, Thermo Scientific™
Chemical Identifiers
CAS | 109-43-3 |
---|---|
Molecular Formula | C18H34O4 |
Molecular Weight (g/mol) | 314.46 |
MDL Number | MFCD00027218 |
InChI Key | PYGXAGIECVVIOZ-UHFFFAOYSA-N |
Synonym | butyl sebacate, decanedioic acid, dibutyl ester, di-n-butyl sebacate, dibutyl sebacate, dibutyl sebacinate, kodaflex dbs, monoplex dbs, polycizer dbs, sebacic acid, dibutyl ester, staflex dbs |
PubChem CID | 7986 |
IUPAC Name | dibutyl decanedioate |
SMILES | CCCCOC(=O)CCCCCCCCC(=O)OCCCC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC406652500
|
Thermo Fisher Scientific
406652500 |
250g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC406650010
|
Thermo Fisher Scientific
406650010 |
1kg | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC406650025
|
Thermo Fisher Scientific
AC406650025 |
2.5kg | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
109-43-3 | |
314.46 | |
PYGXAGIECVVIOZ-UHFFFAOYSA-N | |
7986 | |
CCCCOC(=O)CCCCCCCCC(=O)OCCCC |
C18H34O4 | |
MFCD00027218 | |
butyl sebacate, decanedioic acid, dibutyl ester, di-n-butyl sebacate, dibutyl sebacate, dibutyl sebacinate, kodaflex dbs, monoplex dbs, polycizer dbs, sebacic acid, dibutyl ester, staflex dbs | |
dibutyl decanedioate |
Specifications
109-43-3 | |
Undesignated | |
167°C | |
Authentic | |
CH3(CH2)3OCO(CH2)8COO(CH2)3CH3 | |
Glass bottle | |
butyl sebacate, decanedioic acid, dibutyl ester, di-n-butyl sebacate, dibutyl sebacate, dibutyl sebacinate, kodaflex dbs, monoplex dbs, polycizer dbs, sebacic acid, dibutyl ester, staflex dbs | |
PYGXAGIECVVIOZ-UHFFFAOYSA-N | |
CCCCOC(=O)CCCCCCCCC(=O)OCCCC | |
314.46 | |
314.46 | |
93% | |
Dibutyl sebacate |
0.9400g/mL | |
-12°C | |
91% min. (GC) | |
C18H34O4 | |
MFCD00027218 | |
1.4400 to 1.442 | |
250g | |
0.94 | |
dibutyl decanedioate | |
7986 | |
Liquid | |
0.1% max. (K.F.) |
Safety and Handling
EINECSNumber : 203-672-5
RUO – Research Use Only