missing translation for 'onlineSavingsMsg'
Learn More

Dibromomethane, 99%

CAS: 74-95-3 | CH2Br2 | 173.83 g/mol

Supplier:  Thermo Scientific Chemicals 112830010

 View more versions of this product

Catalog No. AC112830010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful to aquatic life with long lasting effects.
Harmful if inhaled.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

EINECSNumber : 200-824-2

RTECSNumber : PA7350000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only