missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Non-sterile Deep Well Plate for Storage and Collection

Deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage.

$207.40 - $207.40

Specifications

Lid Without Lid
Color Clear
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
View More Specs

Products 1
Catalog Number Mfr. No. Well Shape Total Volume (Metric) Well Price Quantity  
Catalog Number Mfr. No. Well Shape Total Volume (Metric) Well Price Quantity  
10-320-177
View Documents
Corning™
3866
Round 2 mL
Case of 50 Each for $207.40
Only null left
 
Description

Description

  • Made of polypropylene
  • Offer excellent chemical resistance
  • Can withstand temperatures down to -80°C
Specifications

Specifications

Without Lid
Polypropylene
Non-sterile
Translucent
Clear
96
Non-treated
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'