missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Valine methyleester hydrochloride, 99%, Thermo Scientificâ„¢
Chemical Identifiers
CAS | 7146-15-8 |
---|---|
Molecular Formula | C6H13ClNO2 |
Molecular Weight (g/mol) | 166.63 |
MDL Number | MFCD00237309 |
InChI Key | RFMIEHOYULVMNZ-UHFFFAOYNA-N |
Synonym | d-valine methyl ester hydrochloride, methyl d-valinate hydrochloride, h-d-val-ome.hcl, r-methyl 2-amino-3-methylbutanoate hydrochloride, d-valinemethyl ester hydrochloride, methyl 2r-2-amino-3-methylbutanoate hydrochloride, d-valine, methyl ester, hydrochloride, h-d-val-ome, methyl d-valinate hcl, h-d-val-ome hcl |
PubChem CID | 11984198 |
IUPAC Name | methyl (2R)-2-amino-3-methylbutanoate;hydrochloride |
SMILES | [Cl].COC(=O)C(N)C(C)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC398720010
|
Thermo Scientific Chemicals
398720010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC398720050
|
Thermo Scientific Chemicals
398720050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
7146-15-8 | |
166.63 | |
RFMIEHOYULVMNZ-UHFFFAOYNA-N | |
11984198 | |
[Cl].COC(=O)C(N)C(C)C |
C6H13ClNO2 | |
MFCD00237309 | |
d-valine methyl ester hydrochloride, methyl d-valinate hydrochloride, h-d-val-ome.hcl, r-methyl 2-amino-3-methylbutanoate hydrochloride, d-valinemethyl ester hydrochloride, methyl 2r-2-amino-3-methylbutanoate hydrochloride, d-valine, methyl ester, hydrochloride, h-d-val-ome, methyl d-valinate hcl, h-d-val-ome hcl | |
methyl (2R)-2-amino-3-methylbutanoate;hydrochloride |
Specifications
7146-15-8 | |
99% | |
Glass bottle | |
MFCD00237309 | |
d-valine methyl ester hydrochloride, methyl d-valinate hydrochloride, h-d-val-ome.hcl, r-methyl 2-amino-3-methylbutanoate hydrochloride, d-valinemethyl ester hydrochloride, methyl 2r-2-amino-3-methylbutanoate hydrochloride, d-valine, methyl ester, hydrochloride, h-d-val-ome, methyl d-valinate hcl, h-d-val-ome hcl | |
[Cl].COC(=O)C(N)C(C)C | |
1g | |
11984198 | |
≥98.5% |
Authentic | |
C6H13ClNO2 | |
(CH3)2CHCH(NH2)COOCH3·HCl | |
-13.5° to -17.5° (20°C, 589nm) (c=2, H2O) | |
RFMIEHOYULVMNZ-UHFFFAOYNA-N | |
methyl (2R)-2-amino-3-methylbutanoate;hydrochloride | |
166.63 | |
167.63 | |
D-Valine methyleester hydrochloride |
RUO – Research Use Only