missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Tagatose 98.0+%, TCI Americaâ„¢
$168.19 - $548.03
Chemical Identifiers
CAS | 87-81-0 |
---|---|
Molecular Formula | C6H12O6 |
Molecular Weight (g/mol) | 180.156 |
MDL Number | MFCD00134449 |
InChI Key | BJHIKXHVCXFQLS-PQLUHFTBSA-N |
Synonym | 3s,4s,5r-1,3,4,5,6-pentahydroxyhexan-2-one, lyxo-2-hexulose, tagatose, d, d--tagatose, keto-d-tagatose, tagatose nf, unii-t7a20y888y, d-lyxo-hex-2-ulose, naturlose |
PubChem CID | 92092 |
ChEBI | CHEBI:47693 |
IUPAC Name | (3S,4S,5R)-1,3,4,5,6-pentahydroxyhexan-2-one |
SMILES | C(C(C(C(C(=O)CO)O)O)O)O |
Chemical Identifiers
87-81-0 | |
180.156 | |
BJHIKXHVCXFQLS-PQLUHFTBSA-N | |
92092 | |
(3S,4S,5R)-1,3,4,5,6-pentahydroxyhexan-2-one |
C6H12O6 | |
MFCD00134449 | |
3s,4s,5r-1,3,4,5,6-pentahydroxyhexan-2-one, lyxo-2-hexulose, tagatose, d, d--tagatose, keto-d-tagatose, tagatose nf, unii-t7a20y888y, d-lyxo-hex-2-ulose, naturlose | |
CHEBI:47693 | |
C(C(C(C(C(=O)CO)O)O)O)O |
Specifications
87-81-0 | |
White | |
MFCD00134449 | |
3s,4s,5r-1,3,4,5,6-pentahydroxyhexan-2-one, lyxo-2-hexulose, tagatose, d, d--tagatose, keto-d-tagatose, tagatose nf, unii-t7a20y888y, d-lyxo-hex-2-ulose, naturlose | |
C(C(C(C(C(=O)CO)O)O)O)O | |
180.156 | |
CHEBI:47693 | |
≥98.0% (HPLC) | |
D-Tagatose |
132°C | |
C6H12O6 | |
1 g | |
BJHIKXHVCXFQLS-PQLUHFTBSA-N | |
(3S,4S,5R)-1,3,4,5,6-pentahydroxyhexan-2-one | |
92092 | |
180.16 | |
Crystalline Powder |
Safety and Handling
RTECSNumber : WW1100000
TSCA : No