missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D(-)-Fructose, specified according the requirements of USP
CAS: 57-48-7 | C6H12O6 | 180.16 g/mol
Supplier: Thermo Scientific Chemicals 451960010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
D(-)-Fructose | |
103.0°C to 105.0°C | |
C6H12O6 | |
15,4302 | |
d---fructose, d--fructose, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, furucton, arabino-hexulose, keto-d-fructose, d-levulose, sugar, fruit, fructose, d, krystar 300 | |
BJHIKXHVCXFQLS-UYFOZJQFSA-N | |
(3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one | |
5984 | |
180.16 |
57-48-7 | |
0.5% max | |
1 kg | |
− 92.25 | |
Solubility in water: 3750g/L (20°C). Other solubilities: soluble in alcohol,pyridine,ether,ethylamine,and methylamine | |
C(C(C(C(C(=O)CO)O)O)O)O | |
180.16 | |
CHEBI:48095 |
Chemical Identifiers
57-48-7 | |
180.16 | |
d---fructose, d--fructose, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, furucton, arabino-hexulose, keto-d-fructose, d-levulose, sugar, fruit, fructose, d, krystar 300 | |
CHEBI:48095 | |
C(C(C(C(C(=O)CO)O)O)O)O |
C6H12O6 | |
BJHIKXHVCXFQLS-UYFOZJQFSA-N | |
5984 | |
(3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one |
Safety and Handling
EINECSNumber : 200-333-3
RUO – Research Use Only