Learn More
Thermo Scientific™ Cytospin™ 4 Cytocentrifuge Sealed Head Rotor
Use these accessories with the Thermo Scientific™ Cytospin™ 4 Cytocentrifuge.
Supplier:  Thermo Scientific™ 59910018
Catalog No. SH59910018
Only null left
Explore available promotions
Description
Description
Use these accessories with the Thermo Scientific™ Cytospin™ 4 Cytocentrifuge.
  • Shorten preparation time by having a second sealed head ready when the first finishes processing
  • Autoclavable
  • 12-chamber head seals completely
Product Suggestions
Product Suggestions
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'