missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytochrome C From Horse Heart MP Biomedicals
Manufacturer: Mp Biomedicals 1014675
Specifications
Cytochrome C | |
From Horse Heart | |
C42H52FeN8O6S2 | |
WFVBWSTZNVJEAY-UHFFFAOYSA-L | |
3-[8,13-bis[1-[2-amino-3-(methylamino)-3-oxopropyl]sulfanylethyl]-18-(2-carboxyethyl)-3,7,12,17-tetramethylporphyrin-21,24-diid-2-yl]propanoic acid;iron(2+) | |
16057918 |
500g | |
9007-43-6 | |
3-7,12-bis 1-2-amino-3-methylamino-3-oxopropyl sulfanylethyl-18-2-carboxyethyl-3,8,13,17-tetramethylporphyrin-21,23-diid-2-yl propanoic acid; iron 2+ | |
CC1=C(C2=CC3=C(C(=C([N-]3)C=C4C(=C(C(=N4)C=C5C(=C(C(=N5)C=C1[N-]2)C)C(C)SCC(C(=O)NC)N)C)C(C)SCC(C(=O)NC)N)C)CCC(=O)O)CCC(=O)O.[Fe+2] | |
884.893 | |
12000 |
Chemical Identifiers
9007-43-6 | |
884.893 | |
3-7,12-bis 1-2-amino-3-methylamino-3-oxopropyl sulfanylethyl-18-2-carboxyethyl-3,8,13,17-tetramethylporphyrin-21,23-diid-2-yl propanoic acid; iron 2+ | |
3-[8,13-bis[1-[2-amino-3-(methylamino)-3-oxopropyl]sulfanylethyl]-18-(2-carboxyethyl)-3,7,12,17-tetramethylporphyrin-21,24-diid-2-yl]propanoic acid;iron(2+) |
C42H52FeN8O6S2 | |
WFVBWSTZNVJEAY-UHFFFAOYSA-L | |
16057918 | |
CC1=C(C2=CC3=C(C(=C([N-]3)C=C4C(=C(C(=N4)C=C5C(=C(C(=N5)C=C1[N-]2)C)C(C)SCC(C(=O)NC)N)C)C(C)SCC(C(=O)NC)N)C)CCC(=O)O)CCC(=O)O.[Fe+2] |