missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Cyclodextrin hydrate, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 227280050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chelation/Complexation reagentGreener Choice Claims
- Environmental benefits include:
- Recyclable
Specifications
β-Cyclodextrin hydrate | |
68168-23-0 | |
100.0 | |
98.5% min. (ELSD) (HPLC) | |
C42H72O36 | |
5g | |
14, 2718 | |
beta-cyclodextrin hydrate, schardinger, a-dextrin hydrate, beta-cyclodextrin hydrate, puriss hplc | |
5ppm toluene max. | |
O.OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O | |
1153.00 | |
1135.01 | |
1% max. Alpha-and gamma-cyclodextrin (HPLC) |
99% | |
98.5 | |
Authentic | |
Glass bottle | |
MFCD00150811 | |
06,151; 08,133; 09,129; 12,151; 15,107 | |
+ 161 | |
(1% in water) Clear colorless | |
JNSAKCOAFBFODP-UHFFFAOYNA-N | |
5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,â¶.2â¸,¹¹.2¹³,¹â¶.2¹â¸,²¹.2²³,²â¶.2²â¸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol hydrate | |
53486154 | |
99% |
Safety and Handling
TSCA : TSCA
RUO â Research Use Only