missing translation for 'onlineSavingsMsg'
Learn More

Creatine monohydrate, 99%

Creatine monohydrate, CAS # 6020-87-7, is the monohydrate form of creatine, an amino acid found primarily in skeletal muscle cells that is important for rapid energy replenishment. | CAS: 6020-87-7 | C4H9N3O2·H2O | 149.15 g/mol

Supplier:  Thermo Scientific Chemicals 226790250

 View more versions of this product

Catalog No. AC226790250


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Creatine is an amino acid made naturally by the body that can also be acquired through dietary consumption
  • It is primarily stored in skeletal muscle tissue
  • Creatine is stored in a phosphate form, and upon depletion of ATP deposits, phosphocreatine is able to donate phosphate groups to rapidly replenish ATP
  • Creatine supplementation may be beneficial as a disease treatment, particularly in diseases of muscle wasting including cancer and chronic disease

Applications

  • Creatine may be used in the laboratory to study its potential for use as a treatment for metabolic and muscle wasting diseases
  • Creatine has been found to have antitumor, antiviral, and antidiabetic effects
  • It is also used in laboratory settings to study neurological disorders and has been found to be neuroprotective against hypoxic conditions
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only