Learn More
Mopec Creased Dissection Table without Drawers
Designed for the dissection of cadavers and offers a broad dissection area. Mopec™ Creased Dissection Table without Drawers promotes easier transfer of bodies on and off the table.
Supplier:  Mopec HA210
Catalog No. 22444095
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • All stainless steel construction
  • Creased non-removable top
  • Built-in tapered slope
  • Includes drain hole PVC drain valve
  • Four heavy duty 5 in. casters with 3 total locks and 1 directional lock
  • 4 Post sockets included
Warranty
1 Year Warranty
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Specifications
Mobile Dissection Table
30 in.
170.1 kg
Stainless Steel
375 lb.
76.2 cm
Product Suggestions
Product Suggestions