missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Costar™ Clear Multiple Well Plates

Untreated surface is hydrophobic in nature and binds bio-olecules through passive interactions. Corning™ Costar™ Clear Multiple Well Plates provide an ideal solution for immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface.

Supplier:  Corning™ 3736

 View more versions of this product

Catalog No. 07-201-588


Only null left
Explore available promotions

Description

Description

  • Individual alphanumerical codes for well identification, flat bottoms
  • Treated for optimal cell attachment (except where noted)
  • Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
  • Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
  • Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
  • Sterile
  • Nonpyrogenic
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications

Specifications

Standard
9.5 cm2
Medium Binding
Polystyrene
6
Sterile
Flat Bottom (F-well)
5/Pk.
With Lid
2 x 3
Clear
16.8 mL
1.9 to 2.9 mL
Non-treated
100/Cs.
Clear
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions