Learn More
Corning™ Surface Modified Stripwell™ Microplates
Four surface chemistries suit every type of assay
Supplier:  Corning™ 2508
 View more versions of this product
Catalog No. 07-200-142
Only null left
Explore available promotions
Description
Description
Corning™ Stripwell™ microplates may be used in a variety of laboratory assays. The flat bottom strips are designed to easily break apart and are pre-assembled in an “egg-crate” style strip holder that allows each individual well to be positioned back into the microplate once broken.
  • Stripwell microplates have 96-well flat bottom polystyrene format.
  • Low volume and standard Stripwell microplates have well volumes of 190 ÎĽL and 360 ÎĽL, respectively.
  • 1 x 8 strips are designed to fit only one way into the strip holder, eliminating the chance of misorientation.
Specifications
Specifications
Strip Plate
8 x 12
Polystyrene
75 to 200 ÎĽL
Carbo-Bind
Round, Flat Bottom (F-well)
50/Cs.
Without Lid
Clear
96
Non-sterile
360 ÎĽL
In-vitro diagnostic assays
Product Suggestions
Product Suggestions