Learn More
Corning™ Microvolume Pipet Tips, Nonsterile
Provide accurate, reliable performance in the stated volume range
Supplier:  Corning™ C4840
 View more versions of this product
Catalog No. 07-200-280
Only null left
Explore available promotions
Description
Description
  • Microvolume tips provide accurate, reliable performance in the 0.1 ÎĽL to 10 ÎĽL range for major brand ultra-micropipettors.
  • RNase-/DNase-free.
  • Nonpyrogenic.
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Specifications
0.2 to 10 ÎĽL
Natural
Bulk
Bulk
Polypropylene
Non-sterile
DNase, RNase, and Pyrogen free
Microvolume, Round
Gilson™ Pipetman™, Universal Fit Ultramicro
Product Suggestions
Product Suggestions