Learn More
Corning™ LSE™ Vortex Mixer Accessory, Processing Heads
GREENER_CHOICE
Heads allow for processing of larger numbers of tubes
$145.32 - $185.92
Specifications
For Use With (Equipment) LSE Vortex Mixer
View More Specs
Products 5
Catalog Number Mfr. No. Holds Price Quantity  
Catalog Number Mfr. No. Holds Price Quantity  
07-201-794
GREENER_CHOICE
View Documents
Corning™
480100
Multiple Tubes
Case for $145.32
Only null left
07-201-795
GREENER_CHOICE
View Documents
Corning™
480101
24 x 1.5/2.0 mL, 24 x 0.5 mL, 32 x 0.2 mL Tubes/4 Tube Strips
Case for $148.25
Only null left
07-201-796
GREENER_CHOICE
View Documents
Corning™
480102
1 Microplate, 64 x 0.2 mL Tubes, 8 x 0.2 mL Tube Strips
Case for $174.70
Only null left
07-201-797
GREENER_CHOICE
View Documents
Corning™
480103
8 x 15 mL, 8 x 12/13 mm dia. Tubes
Case for $174.70
Only null left
07-201-798
GREENER_CHOICE
View Documents
Corning™
480104
6 x 50 mL Tubes
Case for $185.92
Only null left
Description
Description
This is an optional head for processing larger numbers of tubes, for use with Corning™ LSETM Vortex Mixer.
Specifications
Specifications
LSE Vortex Mixer
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'