missing translation for 'onlineSavingsMsg'
Learn More

Corning™ DeckWorks™ Low-Binding Hinged Rack Pipet Tips, Nonsterile
GREENER_CHOICE

Designed for quantitative assays in which sample binding can significantly alter results.

$262.04 - $1088.09

Specifications

Sterility Non-sterile
Color Natural
For Use With (Application) Designed for quantitative assays in which sample binding can significantly alter results
Format Hinged Rack
Tip Style Low Retention, Round
View More Specs

Products 3
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
10-320-734
View Documents
Corning™
4147
0.1 to 10 ÎĽL
Case of 3840 Each for $1,036.22
Only null left
 
10-320-736
View Documents
Corning™
4149
1 to 300 ÎĽL
Case of 3840 Each for $1,088.09
Only null left
 
10-320-735
View Documents
Corning™
4148
1 to 200 ÎĽL
Pack of 960 Each for $262.04
Case of 4 Pack for $1,048.16
Only null left
 
Description

Description

  • Corning DeckWorks low binding tips are available in 10 ÎĽL, 200 ÎĽL, and 300 ÎĽL sizes.
  • Proprietary low binding technology reduces surface tension on the interior wall of the tip.
  • Leaching and/or subsequent sample degradation associated with silicone-based tips are eliminated.
  • DeckWorks low binding tips are ideal for quantitative assays in which sample binding can significantly alter results.
  • Specific applications include accurate and precise pipetting of expensive reagents, DNA, proteins and/or peptides.
Specifications

Specifications

Non-sterile
Designed for quantitative assays in which sample binding can significantly alter results
Low Retention, Round
Universal Fit
DeckWorks
Natural
Hinged Rack
Hinged rack
Yes
Polypropylene
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'