Learn More
Corning™ DeckWorks™ Hinged Rack Barrier Tips
Features an inert, high-density polyethylene barrier to eliminate aerosol carryover contamination during critical pipetting procedures.
Supplier: Corning™ C4140
Description
- The hinged rack design allows easy rack stacking
- Racks can be reused
- Simply remove the tip insert in the rack and replace with the Corning tip inserts for hinged racks
- Allow maximize storage space and help reduce the production and waste of plastic materials
- Hinged racks within this system are manufactured from recycled polypropylene
- Used for quantitative assays in which sample binding can significantly alter results
- These sterile tips will not trap liquids or inhibit PCR in the event of over-pipetting
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
- Ozone-Safe or Safe for the Ozone Layer
- Free-of a substance that causes environmental harm
- Non-Toxic
Specifications
100 to 1000 ÎĽL | |
Sterile | |
Nonpyrogenic, Human DNA-free, RNase-/DNase-free, Sterility Assurance Level 10-3 | |
Hinged Rack | |
96 Tips/Rack, 8 Racks/Pack | |
DeckWorks |
Filtered | |
Natural | |
Used for quantitative assays in which sample binding can significantly alter results | |
Round | |
Yes | |
HDPE |