missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II,III) oxide, nanopowder, 99% (metals basis)

CAS: 1308-06-1 | Co3O4 | 240.80 g/mol

Supplier:  Thermo Scientific Chemicals 04466136

 View more versions of this product

Catalog No. AA4466136

Only null left
Explore available promotions

Description

Description

Cobalt(II,III) oxide is used in enamels, pigments, porcelain colorants, semiconductors, ceramics, catalysis, grinding wheels and other fields as an important inorganic material. It is a useful oxidant in chemical industry and as catalyst for organic synthesis. It is widely used as catalyst carrier and as electrode active material. It is also used in senior goggles, other filter materials, in temperature and gas sensors, in electronic ceramics, in lithium ion battery electrode materials, magnetic materials, electrochromic devices and solar energy absorbers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.

P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c

H317-H334-H350i

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 215-157-2

RTECSNumber : GG2500000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only